Order Entry
Italy
ContactUsLinkComponent
Pycnometer 10 cm adjusted to third decimal with thermometer and blue filling. Comes with works-certificate. 1 * 1 items
Catalog # HECH91257248316
New Product
Pycnometer 10 cm adjusted to third decimal with thermometer and blue filling. Comes with works-certificate. 1 * 1 items
Catalog # HECH91257248316