Order Entry
Italy
ContactUsLinkComponent
Paliperidone Related Compound E (15 mg) (3-{2-[4-(6-Fluorobenzo[d]isoxazol-3-yl)piperidin-1-yl]ethyl}-2-methyl-7,8-dihydro-4H-pyrido [1,2-a]pyrimidine-4,9(6H)-dione trifluoroacetate) 1 * 15 mg
Catalog # 1491853.
Supplier:  USP
New Product
Paliperidone Related Compound E (15 mg) (3-{2-[4-(6-Fluorobenzo[d]isoxazol-3-yl)piperidin-1-yl]ethyl}-2-methyl-7,8-dihydro-4H-pyrido [1,2-a]pyrimidine-4,9(6H)-dione trifluoroacetate) 1 * 15 mg
Catalog # 1491853.
Supplier:  USP