Nafcillin System Suitability Mixture (15 mg) (This mixture contains nafcillin sodium and ethoxynaphthoic acid) 1 * 15 mg
1450018.
: USP
New Product
Nafcillin System Suitability Mixture (15 mg) (This mixture contains nafcillin sodium and ethoxynaphthoic acid) 1 * 15 mg
1450018.
: USP
New Product
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection