Order Entry
Italy
ContactUsLinkComponent
4 components: 100mg/l each of Ag As Cd Hg in HNO3 5% 1 * 100 mL
Catalog # CMPLRM113433L1
New Product
4 components: 100mg/l each of Ag As Cd Hg in HNO3 5% 1 * 100 mL
Catalog # CMPLRM113433L1
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.