Human Leukopak
Product Details & Documents
Our highly cell-concentrated Leukopak is a specialised product from leukapheresis using the Spectra Optia system. This advanced technology employs centrifugal mechanisms for continuous blood flow, extracting white blood cells from peripheral blood into a sterile bag with ACD-A anticoagulant. The enriched product includes monocytes, lymphocytes, platelets, plasma, and erythrocytes.
- Leukopak is collected from healthy donors using approved permission forms and procedures
- Collection procedures have been approved by the Institutional Review Board (IRB)
- The product includes donor information and serology testing for: HIV, HBs-AG, HCV
- Additional optional information and testing are available for a small fee
Our offerings encompass Leukopaks sourced from our existing inventory, alongside the provision for prospective collections tailored to specific research inquiries. This versatile range ensures a breadth of options to suit a variety of research requirements, whether drawing from our readily available stock or arranging for custom collections based on unique project needs.
Storage: Prepare cells for extended storage in the vapor phase of liquid nitrogen or thaw them for immediate use. Avoid storing cells in the liquid phase of nitrogen. Short-term storage (less than two weeks) at −80 °C is acceptable but should be minimised for optimal stability. Once thawed, it's crucial to use the samples immediately.
Caution: Not intended for any human or animal diagnostic or therapeutic use.
Specifications
Specifications
- Cell Format:Cryopreserved
- Pk:1
- Single Donor/Pooled:Single donor
- Description:Human leukopak, diseased donor