Order Entry
Italy
Orders LinkContactUsLinkComponent
Human Leukopak

Human Leukopak

Human leukopak, healthy donor, 10B cells (10×10⁹ cells/bag), Fresh, Single donor, 1
Catalog # PRTV11192100100
Supplier:   PROVITRO
Human Leukopak
Catalog # PRTV11192100100
Supplier:  PROVITRO
CAS Number:  
Human Leukopak
Human Leukopak

Human leukopak, healthy donor, 10B cells (10×10⁹ cells/bag), Fresh, Single donor, 1

CAS Number:  
Price
5 480,00 €
Log in for availability
1 * 1 pezzi
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Our highly cell-concentrated Leukopak is a specialised product from leukapheresis using the Spectra Optia system. This advanced technology employs centrifugal mechanisms for continuous blood flow, extracting white blood cells from peripheral blood into a sterile bag with ACD-A anticoagulant. The enriched product includes monocytes, lymphocytes, platelets, plasma, and erythrocytes.

  • Leukopak is collected from healthy donors using approved permission forms and procedures
  • Collection procedures have been approved by the Institutional Review Board (IRB)
  • The product includes donor information and serology testing for: HIV, HBs-AG, HCV
  • Additional optional information and testing are available for a small fee

Our offerings encompass Leukopaks sourced from our existing inventory, alongside the provision for prospective collections tailored to specific research inquiries. This versatile range ensures a breadth of options to suit a variety of research requirements, whether drawing from our readily available stock or arranging for custom collections based on unique project needs.

Storage: Prepare cells for extended storage in the vapor phase of liquid nitrogen or thaw them for immediate use. Avoid storing cells in the liquid phase of nitrogen. Short-term storage (less than two weeks) at −80 °C is acceptable but should be minimised for optimal stability. Once thawed, it's crucial to use the samples immediately.

Caution: Not intended for any human or animal diagnostic or therapeutic use.

Specifications

Specifications

  • Cell Format:
    Fresh
  • Pk:
    1
  • Single Donor/Pooled:
    Single donor
  • Description:
    Human leukopak, healthy donor, 10B cells (10×10⁹ cells/bag)

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.