Order Entry
Italy
Orders LinkContactUsLinkComponent
Human Leukopak

Human Leukopak

Human leukopak, healthy donor, 10B cells (10×10⁹ cells/bag), Cryopreserved, Single donor, 1
Catalog # PRTV12192100100
Supplier:   PROVITRO
Human Leukopak
Catalog # PRTV12192100100
Supplier:  PROVITRO
CAS Number:  
Human Leukopak
Human Leukopak

Human leukopak, healthy donor, 10B cells (10×10⁹ cells/bag), Cryopreserved, Single donor, 1

CAS Number:  
Price
5 130,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Our highly cell-concentrated Leukopak is a specialised product from leukapheresis using the Spectra Optia system. This advanced technology employs centrifugal mechanisms for continuous blood flow, extracting white blood cells from peripheral blood into a sterile bag with ACD-A anticoagulant. The enriched product includes monocytes, lymphocytes, platelets, plasma, and erythrocytes.

  • Leukopak is collected from healthy donors using approved permission forms and procedures
  • Collection procedures have been approved by the Institutional Review Board (IRB)
  • The product includes donor information and serology testing for: HIV, HBs-AG, HCV
  • Additional optional information and testing are available for a small fee

Our offerings encompass Leukopaks sourced from our existing inventory, alongside the provision for prospective collections tailored to specific research inquiries. This versatile range ensures a breadth of options to suit a variety of research requirements, whether drawing from our readily available stock or arranging for custom collections based on unique project needs.

Storage: Prepare cells for extended storage in the vapor phase of liquid nitrogen or thaw them for immediate use. Avoid storing cells in the liquid phase of nitrogen. Short-term storage (less than two weeks) at −80 °C is acceptable but should be minimised for optimal stability. Once thawed, it's crucial to use the samples immediately.

Caution: Not intended for any human or animal diagnostic or therapeutic use.

Specifications

Specifications

  • Cell Format:
    Cryopreserved
  • Pk:
    1
  • Single Donor/Pooled:
    Single donor
  • Description:
    Human leukopak, healthy donor, 10B cells (10×10⁹ cells/bag)

Specifications