TRACE METALS IN DRINKING WATER STANDARD 1 * 100 mL
Catalog # ACSDA19C2N01F.L1
Supplier: CPACHEM
New Product
TRACE METALS IN DRINKING WATER STANDARD 1 * 100 mL
Catalog # ACSDA19C2N01F.L1
Supplier: CPACHEM
New Product
Frequently Bought Together
Accessories for pH/°C meter, handheld
Supplier: TESTO
Thioacetamide 4% in aqueous solution Reag. Ph. Eur. 1089602
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
UV/Visible standards, Supelco®
Supplier: Merck
VWR®, Silicone Rotational viscosity standards
Supplier: VWR Chemicals
ICP multi-element standards, CertiPUR®, Supelco®
Supplier: Merck
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
VWR®, Turbidity standards, formazin
Supplier: VWR Chemicals
1-Propanol, Supelco®
Supplier: Merck