Order Entry
Italy
Orders LinkContactUsLinkComponent
Antibiotic Sensitivity Discs
Levofloxacin, 5 mcg, 50
Catalog # HIMESD216-5X50DS
Supplier:   HIMEDIA
Antibiotic Sensitivity Discs
Catalog # HIMESD216-5X50DS
Supplier:  HIMEDIA
CAS Number:  
Antibiotic Sensitivity Discs
Levofloxacin, 5 mcg, 50
CAS Number:  
Price
19,50 €
Log in for availability
1 * 50 pezzi
Product Details & Documents
Antibiotic sensitivity disc are used to determine the effectivness of an antibiotic against a microorganism as a guide to antibiotic therapy.
The disc consist of a specific concentration of an antibiotic in mcg which is tested against a microorganism of a standard density, swabbed uniformly on an appropriate media following overnight incubation.
The antibiotic is impregnated on a filter paper disc in specified concentration. Both sides of the disc are printed with name and concentration for easy identification.
Specifications
Specifications
  • Pk:
    50
  • Description:
    Levofloxacin, 5 mcg
Specifications