Order Entry
Italy
Orders LinkContactUsLinkComponent
Antibiotic Sensitivity Discs
Antibiotic Sensitivity Discs
Ciprofloxacin, 5 mcg, 50
Codice catalogo HIMESD060-5X50DS
Nome del fornitore:   HIMEDIA
Antibiotic Sensitivity Discs
Codice catalogo HIMESD060-5X50DS
Nome del fornitore:  HIMEDIA
Numero CAS:  
Antibiotic Sensitivity Discs
Ciprofloxacin, 5 mcg, 50
Numero CAS:  
Price
19,50 €
Log in for availability
1 * 50 pezzi
Product Details & Documents
Antibiotic sensitivity disc are used to determine the effectivness of an antibiotic against a microorganism as a guide to antibiotic therapy.
The disc consist of a specific concentration of an antibiotic in mcg which is tested against a microorganism of a standard density, swabbed uniformly on an appropriate media following overnight incubation.
The antibiotic is impregnated on a filter paper disc in specified concentration. Both sides of the disc are printed with name and concentration for easy identification.
Specifications
Specifications
  • Pk:
    50
  • Description:
    Ciprofloxacin, 5 mcg