Order Entry
Italy
Orders LinkContactUsLinkComponent
Screw Cap GL 40
Screw Cap GL 40
GL 40, Black, PP
Catalog # RIXIU8S1001401800
Screw Cap GL 40
Catalog # RIXIU8S1001401800
Screw Cap GL 40
GL 40, Black, PP
Price
751,00 €
Log in for availability
1 * 1.800 pezzi
Product Details & Documents
Screw cap made of black PP with ALU laminated PE foam insert for thread GL 40.
  • Height: 14,4 mm
  • Weight: 6 g
The PP screw caps makes it very popular for packaging food and cosmetics, among other things.
Specifications
  • Thread:
    GL 40
  • Colour:
    Black
  • Material:
    PP