Specification: Appearance: White crystals Content (on dried basis): Min. 99.0% Moisture: Max. 3.0% iron: Max. 3ppm pH (5% aq.sol): 5.0-9.0 Ammonia (as NH3): Max. 100mg/kg 1 * 25 kg
Catalog # 802125.PG
Supplier: Thermo Fisher Scientific