Pen-Fix 4 x 3,8 l 1 * 4 items
Catalog # SHAN6101
Supplier: Epredia
New Product
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Toluene, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
1-Butanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor
n-Hexane, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Triethylamine ≥99.6%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.5% (by GC), HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals