28 components: 100mg/l each of Ag Al As B Ba Ca Co Cr Cu Fe K Mg Mn Mo Na Ni Pb Pt Sb Si Sn Te Ti Tl V W Zn Zr in HNO3 5% CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal la 1 * 1 items
Catalog # ACSDRM111998L1
Supplier: CPACHEM