Order Entry
Italy
Orders LinkContactUsLinkComponent
Additional Top and Bottom Shelves
Additional Top and Bottom Shelves
Top shelf, solid, for cleanroom heavy duty tables, W×D×H: 300×1300×700, fabricated from stainless steel 304 with electropolished finish
Catalog # PALBWX1317
Supplier:   Palbam
Additional Top and Bottom Shelves
Catalog # PALBWX1317
Supplier:  Palbam
Additional Top and Bottom Shelves
Additional Top and Bottom Shelves
Top shelf, solid, for cleanroom heavy duty tables, W×D×H: 300×1300×700, fabricated from stainless steel 304 with electropolished finish
Price
1 290,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Documents
Specifications
  • Description:
    Top shelf, solid, for cleanroom heavy duty tables, W×D×H: 300×1300×700, fabricated from stainless steel 304 with electropolished finish
  • Seq:
    19
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..