VIAL 270ML PP WHITE FLIP-TOP HIGH M12 1 * 120 items
Catalog # 216-4910
Supplier: Capitol Europe
New Product
VIAL 270ML PP WHITE FLIP-TOP HIGH M12 1 * 120 items
Catalog # 216-4910
Supplier: Capitol Europe
New Product
Frequently Bought Together
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR®, Powder packs for determination of chlorine, reagents for photometers
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Boot covers, Tyvek® 500, models POBA/POB0
Supplier: DuPont