Order Entry
Italy
ContactUsLinkComponent
Raman Probe 532 nm with hard anodized aluminum housing, Includes 2-1.5m of 100um and with hard anodized aluminum housing, Includes 2-1.5m of 100um and 200um core optical fiber cables in stainless steel housing with FC/FC connectors 1 * 1 items
Catalog # ASYNASY-C-1178
Supplier:  ASYNT
New Product
Raman Probe 532 nm with hard anodized aluminum housing, Includes 2-1.5m of 100um and with hard anodized aluminum housing, Includes 2-1.5m of 100um and 200um core optical fiber cables in stainless steel housing with FC/FC connectors 1 * 1 items
Catalog # ASYNASY-C-1178
Supplier:  ASYNT