M-Aprons, 0,5 mm
M-Aprons 0,5 mm, 1000×1200 mm, Black
BSNC090-128
: Basan
M-Aprons can be utilised in various fields, such as the chemical industry, healthcare, food processing, and cleaning companies.
All aprons are machine washable up to 60 °C but should not be dry cleaned. They are not suitable for use with heat or open flames. Store them at room temperature and avoid creases by rolling them up or hanging them by the neck straps.
- Colour:Black
- Description:M-Aprons 0,5 mm, 1000×1200 mm
- Seq:03
- Cat. No.:BSNC090-128