Order Entry
Italy
ContactUsLinkComponent
EAU HIPERSOLV CHROMANORM HPLC,, VWRC23595.**,, 824.175 mL ACIDE ORTHOPHOSPHORIQUE 85 % HIPERSOLV C,, VWRC153154D,, 0.825,,mL ACETONITRILE HIPERSOLV CHROMA GRADIENT G,, VWRC83639.**,, 1675,,mL 1 * 2,5 L
Catalog # PRLS90328.320
Supplier:  PROLABO CMP
EAU HIPERSOLV CHROMANORM HPLC,, VWRC23595.**,, 824.175 mL ACIDE ORTHOPHOSPHORIQUE 85 % HIPERSOLV C,, VWRC153154D,, 0.825,,mL ACETONITRILE HIPERSOLV CHROMA GRADIENT G,, VWRC83639.**,, 1675,,mL 1 * 2,5 L
Catalog # PRLS90328.320
Supplier:  PROLABO CMP
Frequently Bought Together