Order Entry
Italy
Orders LinkContactUsLinkComponent
3,3'-Dihydroxybenzidine 98%

3,3'-Dihydroxybenzidine 98%

Glass bottle, 1 g
Catalog # 474450010.
Health
3,3'-Dihydroxybenzidine 98%
Catalog # 474450010.
CAS Number:  2373-98-0
3,3'-Dihydroxybenzidine 98%

Glass bottle, 1 g

CAS Number:  2373-98-0
Price
37,20 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  4,4'-Diamino-3,3'-dihydroxybiphenyl, 3,3'-Dihydroxy-4,4'-diaminobiphenyl, 4,4'-Diamino-[1,1'-biphenyl]-3,3'-diol, Benzidine-3,3'-diol
  • CAS Number:2373-98-0
  • Formula:C₁₂H₁₂N₂O₂
  • MDL Number:MFCD00039149
  • Molecular Weight:216.24 g/mol
  • Storage Temperature:Ambient
  • EINECS:220-190-0
Specifications

Specification Test Results

  • Appearance:
    white to brown powder or crystals
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.