Single Use Pressure Sensor, Non-Sterile, Polysulfone, 1 Sanitary Flange Inlet/Outlet, with 0-60psi NIST certificate 1 * 25 items
Catalog # MFLX19406-70
Nome del fornitore: Avantor Fluid Handling
Single Use Pressure Sensor, Non-Sterile, Polysulfone, 1 Sanitary Flange Inlet/Outlet, with 0-60psi NIST certificate 1 * 25 items
Catalog # MFLX19406-70
Nome del fornitore: Avantor Fluid Handling
Frequently Bought Together
Hose Clamps, Adjustable, Stainless Steel
Nome del fornitore: Avantor Fluid Handling
Masterflex® Adapter Fittings, Male Luer to Hose Barb, Straight, Polypropylene, Avantor®
Nome del fornitore: Avantor Fluid Handling
Masterflex® Adapter Fittings, Female Luer to Hose Barb, Straight, Polycarbonate, Avantor®
Nome del fornitore: Avantor Fluid Handling
Heavy-Duty Tubing Pinch Clamps, Polysulfone
Nome del fornitore: Avantor Fluid Handling
HelixMark® Peristaltic Pump Tubing
Nome del fornitore: Freudenberg Medical Europe GmbH
Masterflex® Adapter Fittings, Female Luer to Hose Barb, Straight, Polypropylene, Avantor®
Nome del fornitore: Avantor Fluid Handling
Saint Gobain® Plastic Luer-to-Sanitary Adapters
Nome del fornitore: Avantor Fluid Handling
Masterflex® Adapter Fittings, Luer to Luer, Nylon, Avantor®
Nome del fornitore: Avantor Fluid Handling
Masterflex® One-Way Luer Check Valve Fittings, Luer, Straight, Avantor®
Nome del fornitore: Avantor Fluid Handling
Masterflex® Luer Stopcock Fittings, Polycarbonate, Avantor®
Nome del fornitore: Avantor Fluid Handling
Masterflex® Union Fittings, Hose Barb, Y-Connector, Polypropylene
Nome del fornitore: Avantor Fluid Handling
Smiths Medical® Luer Stopcock Fittings, Polycarbonate
Nome del fornitore: Avantor Fluid Handling