SARS-CoV-2 (COVID-19) Nucleocapsid Monoclonal Antibody [N010] 1 * 0,1 mg
Catalog # PRSI10-544
Supplier: ProSci Inc.
SARS-CoV-2 (COVID-19) Nucleocapsid Monoclonal Antibody [N010] 1 * 0,1 mg
Catalog # PRSI10-544
Supplier: ProSci Inc.
Frequently Bought Together
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, sterile, polypropylene, Corning®
Supplier: Corning
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Centrifuge Tubes, Conical Bottom, 5 ml
Supplier: VWR Collection