Congelatore Verticale -80C Ultra Slim , 577 litri, 4 ripiani/2controsportelli, coibentazione 90 mm VIP, interno INOX Aisi 304, display HPL Touch-Screen TFT 7 1 * 1 pezzi
Catalog # MSPPK62US-HPL
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together