Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Ploxamer 171 (Genapol PF 10) 1 * 200 kg
Catalog # MSPP1000002439
Supplier:
MARKET SOURCE PART PROCESS
Expand image
Ploxamer 171 (Genapol PF 10) 1 * 200 kg
Catalog # MSPP1000002439
Supplier:
MARKET SOURCE PART PROCESS
Frequently Bought Together
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Supelco® Spectroquant® Test Kits, Sulphate
Supplier: Merck
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck