Order Entry
Italy
Orders LinkContactUsLinkComponent
2,3-Dibromo-5-picoline

2,3-Dibromo-5-picoline

Bottle, 25 g
Codice catalogo APOSOR8978-25G
Corrosive
Toxic
Nome del fornitore:   Apollo Scientific
2,3-Dibromo-5-picoline
Codice catalogo APOSOR8978-25G
Nome del fornitore:  Apollo Scientific
Numero CAS:  29232-39-1
2,3-Dibromo-5-picoline

Bottle, 25 g

Numero CAS:  29232-39-1
Price
40,60 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2,3-Dibromo-5-methylpyridine
  • CAS Number:29232-39-1
  • Formula:C₆H₅Br₂N
  • MDL Number:MFCD04112574
  • Molecular Weight:250.92 g/mol
  • Boiling Pt:270.8 °C
  • Melting Pt:52…56 °C
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.