Order Entry
Italy
Orders LinkContactUsLinkComponent
1,1-Dimethoxyacetone

1,1-Dimethoxyacetone

Bottle, 500 g
Catalog # APOSOR40044-500G
Flammable
Supplier:   Apollo Scientific
1,1-Dimethoxyacetone
Catalog # APOSOR40044-500G
Supplier:  Apollo Scientific
CAS Number:  6342-56-9
1,1-Dimethoxyacetone

Bottle, 500 g

CAS Number:  6342-56-9
Price
98,40 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Pyruvaldehyde 1,1-dimethyl acetal, 1,1-Dimethoxypropan-2-one, Pyruvic aldehyde dimethyl acetal
  • CAS Number:6342-56-9
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD00008758
  • Molecular Weight:118.13 g/mol
  • Boiling Pt:133…135 °C
  • Melting Pt:–57 °C
  • Density:1 g/cm³ (25 °C)
  • Flash Pt:37 °C
  • Storage Temperature:Ambient
  • UN:1224
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.