Order Entry
Italy
Orders LinkContactUsLinkComponent
Nile red

Nile red

Bottle, 5 g
Catalog # APOSBIN0465-5G
Supplier:   Apollo Scientific
Nile red
Catalog # APOSBIN0465-5G
Supplier:  Apollo Scientific
CAS Number:  7385-67-3
Nile red

Bottle, 5 g

CAS Number:  7385-67-3
Price
652,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one
  • CAS Number:7385-67-3
  • Formula:C₂₀H₁₈N₂O₂
  • MDL Number:MFCD00011639
  • Molecular Weight:318.38 g/mol
  • Melting Pt:203…205 °C
  • Storage Temperature:Ambient

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications