Cal-520FF™ AM
Plastic bottle, 500 µG
Catalog # AATB21143
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
Cal-520® provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.
- Improved background/signal ratio
- Suitable for all major fluorescence platforms
- Cell permeant
: For research use only.
- Molecular Weight:1138.92 g/mol
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:500 µG
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



