Order Entry
Italy
Orders LinkContactUsLinkComponent
Cal-520FF™ AM

Cal-520FF™ AM

Plastic bottle, 1 mg
Catalog # AATB21142
Irritant
Supplier:   AAT Bioquest
Cal-520FF™ AM
Catalog # AATB21142
Supplier:  AAT Bioquest
CAS Number:  
Cal-520FF™ AM
Cal-520FF™ AM

Plastic bottle, 1 mg

CAS Number:  
Price
785,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Cal-520® provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.

  • Improved background/signal ratio
  • Suitable for all major fluorescence platforms
  • Cell permeant

Caution: For research use only.

  • Molecular Weight:1138.92 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.