Order Entry
Italy
Orders LinkContactUsLinkComponent
Propidium Iodide ≥95%

Propidium Iodide ≥95%

Plastic bottle, 5 g
Catalog # AATB17516
Irritant
Supplier:   AAT Bioquest
Propidium Iodide ≥95%
Catalog # AATB17516
Supplier:  AAT Bioquest
CAS Number:  25535-16-4
Propidium Iodide ≥95%
Propidium Iodide ≥95%

Plastic bottle, 5 g

CAS Number:  25535-16-4
Price
6 240,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Propidium iodide (PI) belongs to the same chemical class of ethidium bromide.

Synonyms:  3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
  • CAS Number:25535-16-4
  • Formula:C₂₇H₃₄I₂N₄
  • MDL Number:MFCD00011921
  • Molecular Weight:668.4 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 g

Specifications