Propidium Iodide ≥95%
Plastic bottle, 25 mg
Catalog # AATB17515
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
Propidium iodide (PI) belongs to the same chemical class of ethidium bromide.
Synonyms: 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
- CAS Number:25535-16-4
- Formula:C₂₇H₃₄I₂N₄
- MDL Number:MFCD00011921
- Molecular Weight:668.4 g/mol
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:25 mg



