Order Entry
Italy
ContactUsLinkComponent
IOS W-PLAN OBJECTIVE 50X/0.95 (OIL) 1 * 1 items
Catalog # OPTKM-634.1
Supplier:  OPTIKA
IOS W-PLAN OBJECTIVE 50X/0.95 (OIL) 1 * 1 items
Catalog # OPTKM-634.1
Supplier:  OPTIKA
Frequently Bought Together