Order Entry
Italy
ContactUsLinkComponent
NaCl 1,6 g KCl 0,0298 g NaH2PO4 0,008 g MgCl2 0,0406 g Glucose 0,198 g HEPES 0,238 g NaHCO3 0,2 g CaCl2 0,0588 g H2O QSP 100mL Ajuster le pH à 7,3 + filtration 0,2 1 * 100 mL
Catalog # PRLS90297.180
Supplier:  PROLABO CMP
NaCl 1,6 g KCl 0,0298 g NaH2PO4 0,008 g MgCl2 0,0406 g Glucose 0,198 g HEPES 0,238 g NaHCO3 0,2 g CaCl2 0,0588 g H2O QSP 100mL Ajuster le pH à 7,3 + filtration 0,2 1 * 100 mL
Catalog # PRLS90297.180
Supplier:  PROLABO CMP
Frequently Bought Together