Fetal Bovine Serum (EU Origin) +Fetal Bovine Serum (South America), pH treated 1 * 500 mL
Catalog # S181P-500
Supplier: Biowest
Fetal Bovine Serum (EU Origin) +Fetal Bovine Serum (South America), pH treated 1 * 500 mL
Catalog # S181P-500
Supplier: Biowest
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
VWR®, Breatheable Rayon Film Seals for Biological Cultures
Supplier: VWR Collection
VWR®, Adhesive PCR Film Seals
Supplier: VWR Collection
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
VWR® Labtop Coolers
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Test and centrifuge tubes
Supplier: BOETTGER PAUL
VWR®, Flat Screw Caps for Microcentrifuge Tubes
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Sealing tape for MicroWell™ plates, Nunc™
Supplier: Thermo Fisher Scientific