The Pepsin Solution is intended to be used for proteolytic pretreatment of formalin-fixed, paraffin-embedded specimens in In Situ Hybridization (ISH). 1 * 4 mL
Catalog # ABNOU0538.4ML
Supplier: Abnova
The Pepsin Solution is intended to be used for proteolytic pretreatment of formalin-fixed, paraffin-embedded specimens in In Situ Hybridization (ISH). 1 * 4 mL
Catalog # ABNOU0538.4ML
Supplier: Abnova
Frequently Bought Together
VWR®, Test Tubes without Rim
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Square Bottles, PET
Supplier: LAB3 OWN PRODUCTION
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
Storage plates, 96-well, MASTERBLOCK®
Supplier: Greiner Bio-One
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom, with Flat Screw Cap
Supplier: VWR Collection
Sealing film, Axygen® AxySeal
Supplier: Corning
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
Cell culture flasks, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Sampling Spoons
Supplier: VWR Collection
Multiwell cell culture plates, Falcon®
Supplier: Corning
VWR®, Flat Screw Caps for Microcentrifuge Tubes
Supplier: VWR Collection
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW