Order Entry
Italy
ContactUsLinkComponent
CEN 6 CISH Probe is designed for the qualitative detection of human chromosome 6 alpha satellites in formalin-fixed, paraffin-embedd ed specimens by chromogenic in situ hybridization (CISH). 1 * 400 µl
Catalog # ABNOCC0004.400UL
Supplier:  Abnova
CEN 6 CISH Probe is designed for the qualitative detection of human chromosome 6 alpha satellites in formalin-fixed, paraffin-embedd ed specimens by chromogenic in situ hybridization (CISH). 1 * 400 µl
Catalog # ABNOCC0004.400UL
Supplier:  Abnova
Frequently Bought Together