Order Entry
Italy
ContactUsLinkComponent
T7 RNA Polymerase is a DNA-dependent RNA polymerase from T7 phage, that possesses a strong and specific 5' to 3' RNA polymerase acti vity. T7 RNA Polymerase has high specificity for T7 promoter sequences and will synthesize large quantities of RNA fr 1 * 100 µl
Catalog # ABNOP9367.100UL
Supplier:  Abnova
T7 RNA Polymerase is a DNA-dependent RNA polymerase from T7 phage, that possesses a strong and specific 5' to 3' RNA polymerase acti vity. T7 RNA Polymerase has high specificity for T7 promoter sequences and will synthesize large quantities of RNA fr 1 * 100 µl
Catalog # ABNOP9367.100UL
Supplier:  Abnova
Frequently Bought Together