Order Entry
Italy
ContactUsLinkComponent
Safety barrel 10 ltr. with self-closing tap ¾ , indicator from Teflon Adjustment: with additional thread opening G3 on top incl. screw cap G3 with gasket from EPDM 1 * 1 items
Catalog # ROETSA-10ZI-G3-ORG
Safety barrel 10 ltr. with self-closing tap ¾ , indicator from Teflon Adjustment: with additional thread opening G3 on top incl. screw cap G3 with gasket from EPDM 1 * 1 items
Catalog # ROETSA-10ZI-G3-ORG
Frequently Bought Together