PORTION CUP TRSP. D62 H32MM 2OZ 60ML 1 * 2.500 items
Catalog # KING723062/201
Supplier: KING DISPOSABLES
PORTION CUP TRSP. D62 H32MM 2OZ 60ML 1 * 2.500 items
Catalog # KING723062/201
Supplier: KING DISPOSABLES
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Bottles, wide neck, amber, with GLS 80® thread and screw cap, DURAN®
Supplier: DWK Life Sciences
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Acetone, EMPLURA®, Reagent Grade for synthesis, Supelco®
Supplier: Merck