1-[4-(PROPAN-2-YL)PHENYL]ETHAN-1-OL 1 * 1 items
Catalog # PROOTRCN756768-1G
Supplier: LGC Standards PROMOCHEM
1-[4-(PROPAN-2-YL)PHENYL]ETHAN-1-OL 1 * 1 items
Catalog # PROOTRCN756768-1G
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions in sachets
Supplier: Mettler - Toledo
Reference standard kits for use with MD 100 and MD 200 photometers
Supplier: Lovibond
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
Benzophenone, Supelco®
Supplier: Merck