Order Entry
Italy
ContactUsLinkComponent
Disposal container, Multi-Safe twin plus, 5,000 ml, yellow, with plastic label plastic label, label/print: yellow/red, material: PP, application e.g.: Disposal of sharp and pointed sharp objects, opening: Oval, height: 379 mm, lid (LxW): 179 x 176 mm 1 * 34 items
Catalog # SARS77.3884.050
Supplier:  SARSTEDT
Disposal container, Multi-Safe twin plus, 5,000 ml, yellow, with plastic label plastic label, label/print: yellow/red, material: PP, application e.g.: Disposal of sharp and pointed sharp objects, opening: Oval, height: 379 mm, lid (LxW): 179 x 176 mm 1 * 34 items
Catalog # SARS77.3884.050
Supplier:  SARSTEDT
Frequently Bought Together