Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Nitrobenzene D5 [CAS:4165-60-0] 2-Fluorobiphenyl [CAS:321-60-8] p-Terphenyl D14 [CAS:1718-51-0] in Dichloromethane 1 * 10 Ampoul
Catalog # CMPL4E8C1KDCM1.10
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
Nitrobenzene D5 [CAS:4165-60-0] 2-Fluorobiphenyl [CAS:321-60-8] p-Terphenyl D14 [CAS:1718-51-0] in Dichloromethane 1 * 10 Ampoul
Catalog # CMPL4E8C1KDCM1.10
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
Supplier: Merck
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Sodium hydroxide 10% in aqueous solution, EMSURE® for analysis, Supelco®
Supplier: Merck
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck