Order Entry
Italy
ContactUsLinkComponent
Model technology: Filtration ductless fume hood Captair 391 Smart Filters configuration: 2C Detection: S Additional options: light button Back panel: transparent Front and Side panel: sequential opening+ standard Worksurface: stainless steel Power su 1 * 1 items
Catalog # ERLAAA611B01121D21
Supplier:  ERLAB CAPTAIRE
Model technology: Filtration ductless fume hood Captair 391 Smart Filters configuration: 2C Detection: S Additional options: light button Back panel: transparent Front and Side panel: sequential opening+ standard Worksurface: stainless steel Power su 1 * 1 items
Catalog # ERLAAA611B01121D21
Supplier:  ERLAB CAPTAIRE
Frequently Bought Together