Order Entry
Italy
ContactUsLinkComponent
Moticam 1080N BMH Attachable HDMI c-mount Camer cmos 1/2,8 Still Resolution: 6mp (3264x1836) Live Resolution: Full HD (1920x1080)USB 2.0, HDMI 1080p and Micro-sd Card Slot Motic Images plus 3.1 and Mot 1 * 1 items
Catalog # MOTI1100600101671
Supplier:  MOTIC
undefined
Moticam 1080N BMH Attachable HDMI c-mount Camer cmos 1/2,8 Still Resolution: 6mp (3264x1836) Live Resolution: Full HD (1920x1080)USB 2.0, HDMI 1080p and Micro-sd Card Slot Motic Images plus 3.1 and Mot 1 * 1 items
Catalog # MOTI1100600101671
Supplier:  MOTIC
Frequently Bought Together