IODINE AND POTASSIUM IODIDE TS 1 1 * 100 mL
Catalog # CMPLUSP232.L1
Supplier: CUSTOM MADE CHEMICALS LAB
IODINE AND POTASSIUM IODIDE TS 1 1 * 100 mL
Catalog # CMPLUSP232.L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Total Iron Test Strips, Dosatest, VWR®
Supplier: VWR Chemicals
Potassium hydroxide 85.0-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Test strips, nitrite, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide ≥97.0%, shot, TECHNICAL
Supplier: VWR Chemicals
Sodium hydroxide 32% in aqueous solution, GPR RECTAPUR®, pure for nitrogen analysis
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Test strips for nickel, MQuant®, Supelco®
Supplier: Merck
NANOCOLOR® Photometric Tube Test Kits for Nitrate
Supplier: MACHEREY-NAGEL
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test kits, photometric, Formaldehyde, LCK cuvette tests
Supplier: Hach
Anthrone in ACS Reagent grade water 97%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
Reagents for DEHA determination with Lovibond® MD photometers
Supplier: Lovibond
Bromocresol green sodium salt 0.04% in aqueous solution, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY