Order Entry
Italy
ContactUsLinkComponent
TANGO-2 mini controller • Control of 1 or 2 axes • High position resolution: 819,200 microsteps / revolution • Compact steel case • Dimensions: 29 x 164 x 62 mm • EMV suitable construction • Solid steel case: robust, shielded plug connectors 1 * 1 items
Catalog # LEIM33080000
Supplier:  LEICA MICROSYSTEMS
TANGO-2 mini controller • Control of 1 or 2 axes • High position resolution: 819,200 microsteps / revolution • Compact steel case • Dimensions: 29 x 164 x 62 mm • EMV suitable construction • Solid steel case: robust, shielded plug connectors 1 * 1 items
Catalog # LEIM33080000
Supplier:  LEICA MICROSYSTEMS
Frequently Bought Together