Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
SARBMCA36S3CE12H
:
Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
SARBMCA36S3CE12H
:
Sartorius Balances
Frequently Bought Together
VWR®, Digital Mini Block Heaters
: VWR Collection
Magnetic stirrers, MIXdrive series
: 2MAG
Heating and drying ovens, Universal UF/UFplus (forced convection)
: MEMMERT
Furnace ashing lvt9/11/c550 tMax1100 with liFt Door anD over-teMPerature liMiter anD therMocouPle hole in the Furnace Door 1 * 1 items
: NABERTHERM
Gas burner, powerjet/powerjet 2
: WARTEWIG
Accessories for Binder incubators
: Binder
Furnace MuFFle with scale anD soFtware lt9/12/sw/c550 tMax1200 with liFt Door anD over-teMPerature liMiter anD therMocouPle hole in the rear wall 1 * 1 items
: NABERTHERM
Accessories for incubators Series B, BD, BD, BD-S, BF, BF, E, ED, ED, ED-S, FD, FD, FDL, FD-S, FED, FED, FP, KB, KT, M
: Binder
TENSION BAR Q FOR SM 1 * 1 items
: Edmund Bühler GmbH
Air circulation FurnAce na 250/65 c540 tMAx 650°c 1 * 1 items
: NABERTHERM
Crucible tongs
: NABERTHERM
Carrier for round bottom flasks, IKA®
: IKA