Order Entry
Italy
ContactUsLinkComponent
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D & Activated Draft Shiel d Motors 1 * 1 items
Catalog # SARBMCA36P3CE123M
Supplier:  Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D & Activated Draft Shiel d Motors 1 * 1 items
Catalog # SARBMCA36P3CE123M
Supplier:  Sartorius Balances
Frequently Bought Together