Achiral semi-prep dcpak® pmpc 250x10mm 5 micron 1 * 1 items
DAICAD335
Frequently Bought Together
VWR®, Manual Crimper and Decapper
: VWR Collection
Spillage trays
: S.C.A.T.
VWR®, Short Thread Vials ND9
: VWR Collection
Microvolume syringes 25 to 500 µl
: Trajan Scientific and Medical
Avantor® ACE® C18, HPLC Columns, 5 µm
: Avantor
Aluminium oxide for preparative chromatography
: MERCK PRODUCTION CHEMICALS
Uniguard™ direct connection guard cartridge holders for Accucore™ columns
: Thermo Fisher Scientific
Funnel with ball and lid, ARNOLD, V2.0
: S.C.A.T.
Snap ring vials, N 11
: MACHEREY-NAGEL
Screw caps for high recovery vials, E-Z Ex-Traction™, WHEATON®
: DWK Life Sciences
Canisters, S60/61, with level indicator, b.safe
: Bohlender
VWR®, Screw Caps for Screw Neck Vials ND15
: VWR Collection