Rabbit monoclonal [EPR28366-42] to GABA A Receptor beta 2/GABRB2 1 * 100 µl
Catalog # ABCAAB309540-100UL
Supplier: Abcam
Rabbit monoclonal [EPR28366-42] to GABA A Receptor beta 2/GABRB2 1 * 100 µl
Catalog # ABCAAB309540-100UL
Supplier: Abcam
Frequently Bought Together
VWR®, Square Bottles, PET
Supplier: LAB3 OWN PRODUCTION
Nalgene®, Biotainer® Bottles
Supplier: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
VWR® Cell Culture Dishes
Supplier: VWR Collection
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
Microplates, 96-well
Supplier: RATIOLAB
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Cryoboxes, 133×133 mm
Supplier: RATIOLAB
High Sensitivity DNA ScreenTape Analysis (ScreenTape and Reagents)
Supplier: AGILENT
Ligation Sequencing Kits
Supplier: Oxford Nanopore Technologies
VWR® Adhesive Sealing Strips for 8-Strip PCR Tubes
Supplier: VWR Collection
PCR microtube strips, 0.2 ml, with attached cap strips, Amplitube™
Supplier: Simport Scientific
Lids for microplates, Matrix
Supplier: THERMO MATRIX TECHNOLOGIES
Cap insert for Cryo.s pink 1 * 50 items
Supplier: Greiner Bio-One
Collagen I-coated microplates, Corning® BioCoat®
Supplier: Corning